Search
| Catalog Number | Quantity |
|---|---|
| DR0120M | 120 Tests/Kit |
Use this dry latex test designed to demonstrate by slide agglutination, Escherichia coli strains possessing the O157 serogroup antigen.
The test is best used in conjunction with Sorbitol MacConkey Agar CM0813. E. coli O157:H7 strains do not ferment sorbitol and, therefore, give colorless colonies on this medium. The majority of E. coli isolates do ferment sorbitol and give characteristic pink colonies. Sorbitol MacConkey Agar should be used as the primary screen. Non-sorbitol fermenting colonies can then be tested with the latex reagents to determine whether the isolate belongs to the O157 serogroup and, therefore, a potential verotoxin-producing strain.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
A qualitative latex slide agglutination test for the detection and species identification of Shigella isolates. Used in a diagnostic workflow to aid clinicians in treatment options for patients suspected of having bacterial infections. The device is not automated, is for professional use only and is not a companion diagnostic.
Share catalog number, name or link