Search Thermo Fisher Scientific
Catalog Number | Quantity |
---|---|
DR0130M | 24 Tests/Kit |
Thermo Scientific™ Oxoid Pylori is a rapid latex agglutination screening test is used for qualitative detection of Helicobacter pylori antibodies in serum.
Includes:
8 reagent cards (3 tests/card); Positive Control; Negative Control; Dilution Buffer; mixing sticks; storage pouch
Share catalog number, name or link