Search Thermo Fisher Scientific
Catalog Number | Description | Quantity |
---|---|---|
DR0200M | DrySpot Legionella Pneumophila Serogroup 1 Latex Test | 60 Tests/Kit |
The DrySpot Legionella Latex Test is a latex agglutination test for the identification of Legionella species grown on plate media from patients with suspected legionellosis or from environmental sources. The DrySpot Legionella Latex Test allows separate identification of Legionella pneumophila serogroup 1, serogroups 2–14 and seven other Legionella species that have been implicated in human disease.
A qualitative latex agglutination assay for the confirmatiory identification of legionella pneumophilia serogroup 1 isolates from isolates grown on agar. Used in a diagnostic workflow to aid clinicians in treatment options for patients suspected of having bacterial infections The device is not automated, is for professional use and not is a companion diagnostic.
Share catalog number, name or link