Oxoid™ PBP2' Latex Agglutination Test Kit
Oxoid™ PBP2' Latex Agglutination Test Kit
Thermo Scientific™
Oxoid™ PBP2' Latex Agglutination Test Kit
For detection of PBP2' and identification of methicillin resistance in Staphylococcus aureus (MRSA) and coagulase-negative Staphylococci.
Have Questions?
Catalog NumberQuantity
DR0900A50 Tests/Kit
Catalog number DR0900A
Price (USD)
-
Request A Quote
Quantity:
50 Tests/Kit
Ask our AI about this Product
Thermo Scientific™ Oxoid PBP2' Latex Agglutination Test is a rapid latex slide agglutination test for detection of PBP2' as an aid in identifying methicillin resistance in both Staphylococcus aureus (MRSA) and coagulase-negative Staphylococci.
Ordering Alerts:
50 tests/kit.
Specifications
For Use With (Application)Healthcare
Quantity50 Tests/Kit
Product TypeLatex Agglutination Test Kit
Unit SizeEach
Intended use statement 
A qualitative latex slide agglutination test for the differentiation of meticillin resistant Staphylococcus aureus from other Staphylococcus species isolates by the detection of low affinity penicillin-binding protein PBP2' encoded by the mecA gene from isolates grown on agar. Used in a diagnostic workflow to aid clinicians in treatment options for patients suspected of MRSA infection or colonization. The device is not automated, is for professional use only and is not a companion diagnostic.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Share catalog number, name or link
1x1 image pixel for data collection