Thermo Scientific™
Nalgene™ Polypropylene Validation Bottles, 30 mL
Catalog number: DS2126-0030
Have Questions?
Thermo Scientific™
Nalgene™ Polypropylene Validation Bottles, 30 mL
Catalog number: DS2126-0030
Have Questions?
Catalog Number
DS2126-0030
Unit Size
Case of 30
Price (USD)
Catalog NumberUnit SizePrice (USD)
DS2126-0030Case of 30Request A Quote
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Perform QA lot sample storage of high-valued pharmaceuticals using Thermo Scientific™ Nalgene™ Polypropylene Validation Bottles. Small-volume bottles are molded of the same materials as larger carboys, but require a lower volume of expensive reagents to perform validation testing. Autoclavable, transparent polypropylene construction allows for convenient material compatibility validation. Includes: White polypropylene screw closure and TPE gasket.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Capacity (Metric)
30 mL
Material
Polypropylene
Sterility
Non-sterile
Mouth
Wide
Closure Size
20-415 mm