Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Perform QA lot sample storage of high-valued pharmaceuticals using Thermo Scientific™ Nalgene™ Polycarbonate Validation Bottles. Small-volume bottles are molded of the same materials as larger carboys, but require a lower volume of expensive reagents to perform validation testing.
Autoclavable | Autoclavable |
Capacity (English) | 0.5 gal. |
Capacity (Metric) | 2 L |
Closure Material | Polypropylene |
Closure Size | 53B |
Color | Clear |
Diameter (Metric) Inner Neck | 42 mm |
Diameter (Metric) Outer | 123 mm |
For Use With (Application) | Perform validation testing |
Height (Metric) With Closure | 296 mm |
Material | Polycarbonate |
Mouth Style | Wide |
No. per Case | 12/Cs. |
No. per Pack | 12 Pk. |
Product Type | Validation Bottle |
Sterility | Non-sterile |
Unit Size | Case of 12 |
Autoclavable | Autoclavable |
Capacity (English) | 1 oz. |
Capacity (Metric) | 30 mL |
Closure Material | Polypropylene |
Closure Size | 20-415 |
Color | White (Translucent) |
Diameter (Metric) Inner Neck | 14 mm |
Diameter (Metric) Outer | 32 mm |
For Use With (Application) | Perform validation testing |
Height (Metric) With Closure | 75 mm |
Material | Polycarbonate |
Mouth Style | Wide |
No. per Case | 30/Cs. |
No. per Pack | 30 Pk. |
Product Type | Validation Bottle |
Sterility | Non-sterile |
Unit Size | Case of 30 |
Autoclavable | Autoclavable |
Capacity (English) | 8 oz. |
Capacity (Metric) | 250 mL |
Closure Material | Polypropylene |
Closure Size | 53B |
Color | White (Translucent) |
Diameter (Metric) Inner Neck | 45 mm |
Diameter (Metric) Outer | 74 mm |
For Use With (Application) | Perform validation testing |
Height (Metric) With Closure | 135 mm |
Material | Polycarbonate |
Mouth Style | Wide |
No. per Case | 6/Cs. |
No. per Pack | 6 Pk. |
Product Type | Validation Bottle |
Sterility | Non-sterile |
Unit Size | Case of 6 |
Catalog Number | Specifications | Unit Size | Capacity (Metric) | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
DS2127-2000 | Case of 12 | 2 L | 696.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
DS2127-0030 | Case of 30 | 30 mL | 693.65 Online price 814.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
DS2127-0250 | Case of 6 | 250 mL | 211.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Includes:
White polypropylene screw closure and TPE gasket