Nalgene™ Polypropylene Fluid-Transfer Closure
Nalgene™ Polypropylene Fluid-Transfer Closure
Nalgene™ Polypropylene Fluid-Transfer Closure
Nalgene™ Polypropylene Fluid-Transfer Closure
Thermo Scientific™

Nalgene™ Polypropylene Fluid-Transfer Closure

Perform fluid transfer in the lab or field with this 70mm closure compatible with Thermo Scientific™ Nalgene™ Mason Jars (Cat. No. 2115).
Have Questions?
Catalog NumberThread Size (Metric)
DS2153-0700S2 mm
Catalog number DS2153-0700
Price (USD)
56.56
Online Exclusive
70.70
Save 14.14 (20%)
Case of 6
Thread Size (Metric):
S2 mm
Price (USD)
56.56
Online Exclusive
70.70
Save 14.14 (20%)
Case of 6

Perform fluid transfer in the lab or field with the 70mm Thermo Scientific™ Nalgene™ Polypropylene Fluid-Transfer Closure. Closures are compatible with Nalgene Mason Jars (Cat. No. 2115), 125 and 500mL Nalgene Straight-sided jars (Cat. Nos. 2116, 2117, 2118, 2119), sold separately. Use these closures for siphoning, evacuation, bleeding of fluid lines, collecting samples or transferring reagents. Two vent ports are molded into the top of the closure (external male ports only; no connection for internal drop tubes), allowing for external connection to a vacuum or pressure pump with 1/4 in. I.D. flexible tubing (tubing sold separately).

  • Closure fits on Nalgene Mason Jars or 125 and 250mL straight-sided jars for a perfect fit
  • External port tubes are molded into closure for longer life
  • Vacuum limit: 5″ (127mm) Hg
  • Pressure limit: 2psig (0.137 bar)
  • Autoclavable
Specifications
AutoclavableAutoclavable
DescriptionNalgene Polypropylene Flud-Transfer Closure, 6/cs.
For Use With (Equipment)Nalgene Mason Jars or 125 and 250mL straight-sided jars
Neck Finish70 mm
No. per Case6
No. per Pack6
Thread Size (Metric)S2 mm
ColorWhite
MaterialPolypropylene
Product TypeFluid Transfer Cap
Unit SizeCase of 6