Search Thermo Fisher Scientific
Catalog Number | Type | Includes |
---|---|---|
DT80 | General Purpose Datalogger | Logger and accessories |
DT80M | General Purpose Datalogger | Logger with integrated modem and accessories |
DT80W | General Purpose Datalogger | Logger with integrated Wifi and accessories |
DT80G-AL | Geotechnical logger supporting vibrating wire strain gauges | Logger only (no accessories) |
DT80G | Geotechnical logger supporting vibrating wire strain gauges | Logger and accessories |
DT80-AL | General Purpose Datalogger | Logger only (no accessories) |
The DataTaker DT80 series comprises a family of industrial data loggers, ranging from five channel data loggers to systems with hundreds of analog inputs and many digital inputs.
The DataTaker DT80 series measures and logs analog & digital data from numerous electrical sensors or analog instruments, RTDs, thermocouples, temperature sensors, strain gauges, current loops, voltages, resistance and frequencies.
DataTaker DT80 models include an Ethernet port, USB port and a built-in web server, dEX for setup and for viewing/downloading of data. In addition:
DataTaker DT80G is specifically suited to geotechnical applications such as slope stability, subsidence, dam wall monitoring, tunnel and mining excavation, ground water, tunnel wall monitoring and site assessment. DT80G supports vibrating wire sensors and other geotechnical sensors.
Other Features:
Share catalog number, name or link