Invitrogen™

Eosin-5-Maleimide

Catalog number: E118
Have Questions?
Invitrogen™

Eosin-5-Maleimide

Catalog number: E118
Have Questions?
Catalog Number
E118
Unit Size
25 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
E11825 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Bioconjugates made with the thiol-reactive eosin-5-maleimide can be used as phosphorescent probes or as photosensitizers. With its high quantum yield (∼0.57) for singlet oxygen generation, eosin and its conjugates can be used as effective photooxidizers of diaminobenzidine (DAB) in high-resolution electron microscopy studies and in correlated fluorescence and electron microscopy applications.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Quantity
25 mg
Product Type
Maleimide
Chemical Reactivity
Thiol
Shipping Condition
Room Temperature
Reactive Moiety
Maleimide
Label or Dye
Eosin
Label Type
Classic Dyes

Contents & Storage

Store in freezer (-5 to -30°C) and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text