EGTA, Tetra(acetoxymethyl Ester) (EGTA, AM)
EGTA, Tetra(acetoxymethyl Ester) (EGTA, AM)
Invitrogen™
EGTA, Tetra(acetoxymethyl Ester) (EGTA, AM)
The cell-permeant EGTA, AM can be passively loaded into cells to generate intracellular EGTA. This compound is intrinsically a liquidRead more
Have Questions?
Catalog NumberQuantity
E121910 mg
Catalog number E1219
Price (USD)
524.00
Each
-
Add to cart
Quantity:
10 mg
Price (USD)
524.00
Each
Add to cart
Ask our AI about this Product
The cell-permeant EGTA, AM can be passively loaded into cells to generate intracellular EGTA. This compound is intrinsically a liquid or an oil at room temperature.
Learn more about ion indicators including calcium, potassium, pH, and membrane potential indicators ›
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Dye TypeChelator (Non-Fluorescent)
Quantity10 mg
Shipping ConditionRoom Temperature
Product TypeEGTA AM
Unit SizeEach
Contents & Storage
Store in freezer -5°C to -30°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link