Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | EnzChek Caspase-3 Assay Kit #2, Z-DEVD-R110 substrate |
Format | Cuvets, 96-well Plate |
Product Type | Caspase Assay Kit |
Excitation/Emission | 496/520 (cleaved substrate) |
No. of Reactions | 500 assays (reaction volume 100μL per assay) |
Shipping Condition | Room Temperature |
Product Line | EnzChek |
Detection Method | Fluorescence |
For Use With (Equipment) | Fluorometer, Microplate Reader |
Label or Dye | Rhodamine 110 |
Label Type | Other Label(s) or Dye(s) |
Storage Requirements | Store in freezer (-5 to -30°C) and protect from light. |
Unit Size | 500 assays |
Description | EnzChek Caspase-3 Assay Kit #1, Z-DEVD-AMC substrate |
Format | Cuvets, 96-well Plate |
Product Type | Caspase Assay Kit |
Excitation/Emission | 342/441 (cleaved substrate) |
No. of Reactions | 500 assays (reaction volume 100μL per assay) |
Shipping Condition | Room Temperature |
Product Line | EnzChek |
Detection Method | Fluorescence |
For Use With (Equipment) | Fluorometer, Microplate Reader |
Label or Dye | AMC (7-amino-4-methylcoumarin) |
Label Type | Other Label(s) or Dye(s) |
Storage Requirements | Store in freezer (-5 to -30°C) and protect from light. |
Unit Size | 500 assays |
Catalog Number | Specifications | Unit Size | Description | Label or Dye | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
E13184 | 500 assays | EnzChek Caspase-3 Assay Kit #2, Z-DEVD-R110 substrate | Rhodamine 110 | 560.65 Online price 574.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
E13183 | 500 assays | EnzChek Caspase-3 Assay Kit #1, Z-DEVD-AMC substrate | AMC (7-amino-4-methylcoumarin) | 493.65 Online price 500.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|