Invitrogen™

EKMax™ Enterokinase

Catalog number: E18002
Have Questions?
Invitrogen™

EKMax™ Enterokinase

Catalog number: E18002
Have Questions?
Catalog Number
E18002
Unit Size
1,000 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
250 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
E180021,000 units
-
E18001250 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
EKMax™ is a recombinant preparation of the catalytic subunit of bovine enterokinase (1). EKMax™ recognizes the sequence Asp-Asp-Asp-Asp-Lys and cleaves the peptide bond after the lysine residue. The enzyme can be used to cleave any fusion protein that carries this peptide sequence (Figure 1).

Application: Removal of fusion tags from recombinant proteins.

Unit Definition: One unit of EKMax™ is the amount of enzyme required to digest 20 μg of a thioredoxin-CAT fusion protein to 90% completion in 16 hours at 37°C. One EKMax™ unit is equivalent to ∼190 trypsinogen activation units.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Enterokinase
Species
Cattle/Bovine
Protein Tag
His Tag
Quantity
1000 Units
Product Line
EKMax™

Contents & Storage

EKMax™ Enterokinase is provided with 10X buffer. Store at -20°C. Guaranteed stable for 6 months when stored properly.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text