Escherichia coli (K-12 strain) BioParticles™, fluorescein conjugate
<i>Escherichia coli</i> (K-12 strain) BioParticles&trade;, fluorescein conjugate
Invitrogen™

Escherichia coli (K-12 strain) BioParticles™, fluorescein conjugate

The Molecular Probes™ BioParticles™ product line consists of a series of fluorescently labeled, heat- or chemically killed bacteria and yeastRead more
Have Questions?
Catalog NumberQuantity
E286110 mg
Catalog number E2861
Price (USD)
294.00
Each
In stock
Add to cart
Quantity:
10 mg
Price (USD)
294.00
Each
Add to cart
The Molecular Probes™ BioParticles™ product line consists of a series of fluorescently labeled, heat- or chemically killed bacteria and yeast in a variety of sizes, shapes, and natural antigenicities. These fluorescent BioParticles™ products have been employed to study phagocytosis by fluorescence microscopy, quantitative spectrofluorometry, and flow cytometry.

We offer E. coli (K-12 strain), S. aureus (Wood strain, without protein A) and zymosan (S. cerevisiae) BioParticles™ conjugates covalently labeled with a variety of different fluorophores (special care has been taken to remove free dye after conjugation). Unlike the fluorescence of fluorescein-labeled BioParticles™ conjugates, which is partially quenched in acidic environments, the fluorescence of the Alexa Fluor™, BODIPY™ FL, tetramethylrhodamine and Texas Red™ dye conjugates is uniformly intense over the pH range from 3 to 10.

BioParticles Specifications:
• Label (Ex/Em): Fluorescein (∼494/518 nm)
• Particle: E. coli (K-12 strain)
Opsonizing reagent available


Using BioParticles Products
BioParticles™ conjugates are provided as lyophilized powders. There are approximately 3 x 108 E. coli or S. aureus particles per mg solid and approximately 2 x 107 zymosan particles per mg solid. BioParticles™ conjugates can be reconstituted in the buffer of your choice for use in phagocytosis assays. The fluorescence of BioParticles™ conjugates that are bound to the surface of the cell (but not internalized) can be quenched by ethidium bromide, trypan blue, or other quenchers. In addition to cellular applications, fluorescent BioParticles™ conjugates may be effective as flow cytometry calibration references when sorting bacteria and yeast mutants. These small particles may also be useful references for light scattering studies because their sizes and shapes differ in characteristic ways.

Find More BioParticles™ Products
We offer a large range of dye-labeled and unlabeled E. coli (K-12 strain), S. aureus (Wood strain, without protein A), and zymosan (S. cerevisiae) BioParticles™ products. Find out about these products and their applications by reviewing Probes for Following Receptor Binding and Phagocytosis—Section 16.1 in the Molecular Probes™ Handbook.

For pH-sensitive endocytosis assays, see our pHrodo™ BioParticles™ conjugates.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Dye TypeFITC (Fluorescein)
FormSolid
Quantity10 mg
Shipping ConditionRoom Temperature
SpeciesE. coli
For Use With (Equipment)Fluorescence Microscope
Product LineBioParticles
Product TypeE.coli Conjugate
pH3 to 10
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Frequently asked questions (FAQs)

How are the Escherichia coli (K-12 strain) BioParticles, fluorescein conjugate supplied?

The Escherichia coli (K-12 strain) BioParticles, fluorescein conjugate are supplied as lyophilized powder in a 10 mg.unit size. There are approximately 3 x 10E8 E. coli particles per mg solid.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Are the Invitrogen BioParticles products sterile?

While the bacteria have been attenuated with formaldehyde and alcohol desiccation, the BioParticles products are not considered sterile, and we do not recommend incubation of more than 4 hours. This applies to all of our dye-labeled (pHrodo, Alexa Fluor, etc.) and unlabeled BioParticles products.

What is the type of bond that attaches the dyes to the BioParticles probes?

We use amine-reactive dyes to covalently attach fluorescent dyes to all of our BioParticles probes such as the Escherichia coli (K-12 strain) BioParticles probes, Staphylococcus aureus (Wood strain without protein A) BioParticles, and the Zymosan A (S. cerevisiae) BioParticles probes.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What can the BioParticles product line be used for?

Fluorescent Bioparticles have been employed to study phagocytosis by fluorescence microscopy, quantitative spectrofluorometry, and flow cytometry. We offer E.Coli, S. aureus, and zymosan BioParticles conjugates covalently labeled with a variety of different fluorophores.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Fluorescence spectra

Fluorescence spectra

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection