Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Color | Red, clear |
Description | Contrast MRSA Broth |
Format | Universal |
Manufacturing Origin | UK |
Product Type | Broth |
Type | Broth |
Fill Volume | 3 mL |
Fill Weight | 3.0 - 3.3g |
Packaging Type | Cardboard Container |
Quantity | 10/Pk. |
Shelf Life | 6 Weeks |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Format | Quantity | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
EB1225B | Each | Contrast MRSA Broth | Universal | 10/Pk. | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|