Invitrogen™

Novex™ 4 to 20%, Tris-Glycine, 1.0 mm, Mini Protein Gel, 1-well

Catalog number: EC6021BOX
Have Questions?
Invitrogen™

Novex™ 4 to 20%, Tris-Glycine, 1.0 mm, Mini Protein Gel, 1-well

Catalog number: EC6021BOX
Have Questions?
Catalog Number
EC6021BOX
Unit Size
10 gels (1 box)
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
EC6021BOX10 gels (1 box)
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Novex™ 4–20% Tris-Glycine Mini Gels are polyacrylamide gels based on traditional Laemmli protein electrophoresis. Novex™ Tris-Glycine gels provide reproducible separation of a wide range of proteins into well-resolved bands. Each box contains 10 gels. Features of these gels:

•   Individually packaged gels for optimal experiment design
•   Compatible with most protein standards
•   Usable for native and denaturing protein assays

Choose the right gel format for your experiments
Novex™ Tris-Glycine gels come in a variety of fixed concentrations from 4% to 18%, as well as gradients with ranges of 4–12%, 4–20%, 8–16%, and 10–20%. You can also select between our mini (8 cm x 8 cm) and our midi (8 cm x 13 cm) sizes, as well as multiple well-formats from 1 well to 26 wells.

For best results, most Novex™ Tris-Glycine gels should be stored at 4°C and used within 8 weeks of purchase. Novex™ 16% and 18% Tris-Glycine gels should be used within 4 weeks of purchase.

Run your proteins in native or denatured form

Novex™ Tris-Glycine gels do not contain SDS and can be used to run your proteins in native or in denatured form. For denatured proteins, we recommend using Novex™ Tris-Glycine SDS Sample Buffer and Novex™ Tris-Glycine SDS Running Buffer. For native proteins, we recommend using Novex™ Tris-Glycine Native Sample Buffer and Novex™ Tris-Glycine Native Running Buffer.

For transferring your proteins to membrane, use Novex™ Tris-Glycine Transfer Buffer. Transfers can be done using the Xcell II™ Blot Module or using the iBlot™ Gel Transfer Device.

Related Links

•   Use our search tool to find the Novex™ Tris-Glycine gel that is right for you.
•   See protein migration ranges on Novex™ Tris-Glycine gels.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment)
Mini Gel Tank, XCell SureLock Mini-Cell
Gel Percentage
4 to 20%
Gel Size
Mini
Gel Thickness
1.0 mm
Gel Type
Tris-Glycine
Product Type
Gradient Page
Sample Loading Volume
700 μL
Separation of
Protein
Separation Range
20 to 200 kDa
Separation Type
Denaturing, Native
Shelf Life
8 Weeks
Wells
1-well
Mode of Separation
Molecular Weight
Product Line
Novex™
Quantity
Each
Shipping Condition
Wet Ice
System Type
Novex™
Thickness
1.0 mm
Well Design
1D Well
Length (Metric)
8 cm
Length (Metric) Gel
8 cm
Width (Metric)
8 cm
Width (Metric) Gel
8 cm

Contents & Storage

One box contains 10 gels. Store in refrigerator (2–8°C). Do not freeze. Shelf life is 8 weeks.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text