Invitrogen™

Novex™ 10%, Tris-Glycine, 1.0 mm, Mini Protein Gel, 2D-well

Catalog number: EC6076BOX
Have Questions?
Invitrogen™

Novex™ 10%, Tris-Glycine, 1.0 mm, Mini Protein Gel, 2D-well

Catalog number: EC6076BOX
Have Questions?
Catalog Number
EC6076BOX
Unit Size
10 gels (1 box)
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
EC6076BOX10 gels (1 box)
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Novex™ Tris-Glycine polyacrylamide gel chemistry is based on the Laemmli system (1) with minor modifications for maximum performance in the pre-cast format. These gels do not contain SDS and can therefore be used to accurately separate both native and denatured proteins. Novex™ Tris-Glycine Gels provide reproducible separation of a wide range of proteins into well-resolved bands.

Formulation: Novex™ Tris-Glycine Gels are made with high-purity, strictly quality-controlled reagents: Tris base, HCl, acrylamide, bisacrylamide, TEMED, APS, and highly purified water. They do not contain SDS.

Recommended Buffers: By choosing the appropriate Novex™ pre-mixed buffer, you can create either native, denaturing or reducing running conditions with any Novex™ Tris-Glycine Gel.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment)
Mini Gel Tank, XCell SureLock Mini-Cell
Gel Percentage
10%
Gel Size
Mini
Gel Thickness
1.0 mm
Gel Type
Tris-Glycine
Product Type
Homogeneous Page
Separation of
Protein
Separation Range
30 to 250 kDa
Separation Type
Denaturing, Native
Shelf Life
8 Weeks
Wells
1-well
Mode of Separation
Molecular Weight
Product Line
Novex™
Quantity
Each
Shipping Condition
Wet Ice
System Type
ZOOM™
Thickness
1.0 mm
Well Design
2D Well
Length (Metric)
8 cm
Length (Metric) Gel
8 cm
Width (Metric)
8 cm
Width (Metric) Gel
8 cm

Contents & Storage

One box contains 10 gels. Store in refrigerator (2–8°C). Do not freeze. Shelf life is 8 weeks.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text