Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | Novex TBE-Urea Gels, 15%, 10-well |
For Use With (Equipment) | XCell SureLock Mini-Cell |
Gel Percentage | 15% |
Gel Type | TBE-Urea |
Product Type | TBE Gel |
Separation Range | 10 to 40 bp |
Wells | 10-well |
Quantity | 10 Gels/Box |
Sample Type | Single-stranded DNA Oligos/RNA |
Shipping Condition | Wet Ice |
Unit Size | 1 box |
Description | Novex TBE-Urea Gels, 15%, 12-well |
For Use With (Equipment) | XCell SureLock Mini-Cell |
Gel Percentage | 15% |
Gel Type | TBE-Urea |
Product Type | TBE Gel |
Separation Range | 10 to 40 bp |
Wells | 12-well |
Quantity | 10 Gels/Box |
Sample Type | Single-stranded DNA Oligos/RNA |
Shipping Condition | Wet Ice |
Unit Size | 1 box |
Description | Novex TBE-Urea Gels, 15%, 15-well |
For Use With (Equipment) | XCell SureLock Mini-Cell |
Gel Percentage | 15% |
Gel Type | TBE-Urea |
Product Type | TBE Gel |
Separation Range | 10 to 40 bp |
Wells | 15-well |
Quantity | 10 Gels/Box |
Sample Type | Single-stranded DNA Oligos/RNA |
Shipping Condition | Wet Ice |
Unit Size | 1 box |
Catalog Number | Specifications | Unit Size | Description | Wells | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
EC6885BOX | 1 box | Novex TBE-Urea Gels, 15%, 10-well | 10-well | 209.65 Online price 228.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
EC68852BOX | 1 box | Novex TBE-Urea Gels, 15%, 12-well | 12-well | 209.65 Online price 228.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
EC68855BOX | 1 box | Novex TBE-Urea Gels, 15%, 15-well | 15-well | 204.65 Online price 222.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Novex TBE-Urea Gels can be stained by silver staining, ethidium bromide, and SYBR™ Green staining techniques after electrophoresis. The compact gels are available in 6%, 10%, or 15% formats and are designed to run on the XCell SureLock™ Mini-Cell.
Advantages of TBE-Urea Gels:
• Contains 7M urea for maximum denaturation
• Excellent resolution for fast size and purity confirmations of DNA or RNA oligos
• High resolution of short single-strand oligonucleotides
• Requires ∼10% sample concentration and volume of large or agarose gels
• Accurate and reproducible results
Formulation
Novex TBE-Urea gels are made with high-purity reagents, including Tris base, boric acid, EDTA, acrylamide, bisacrylamide, TEMED, APS, and 7M urea. Strict quality control ensures that gels and buffers are DNase- and RNase-free.
Recommended Buffers
For optimum performance, Novex™ TBE-Urea sample buffers and Novex™ TBE Running Buffer are strongly recommended for use with these gels. For analytical applications, Novex TBE-Urea Sample Buffer is recommended contains urea, the density agent Ficoll™, which yields sharper, straighter bands than conventional density agents, and the tracking dyes Bromophenol Blue and Xylene Cyanol.