Glutathione Peroxidase (GSH-Px) Activity Kit
Glutathione Peroxidase (GSH-Px) Activity Kit
Invitrogen™

Glutathione Peroxidase (GSH-Px) Activity Kit

The Glutathione Peroxidase (GSH-Px) Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of GSH-PxRead more
Promo ImagePromotion
Promo code:RPUZZ25
Stock up on essentials to piece your discovery together
Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzleLearn More
Have Questions?
Catalog NumberQuantity
EEA010Promo Image96 tests
Catalog number EEA010
Price (USD)
696.00
Each
Add to cart
Quantity:
96 tests
Recurring order eligible. Learn more »
Price (USD)
696.00
Each
Add to cart
Ask our AI about this Product

The Glutathione Peroxidase (GSH-Px) Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of GSH-Px activity in various sample types. GSH-Px is a crucial enzyme that plays a key role in protecting cells against oxidative damage by catalyzing the reduction of hydrogen peroxide and organic hydroperoxides using glutathione as a cofactor, protecting cell membrane structure and keeping membrane function integrity.

The activity of glutathione can be calculated by measuring the consumption of reduced glutathione. GSH can react with dinitrobenzoic acid to produce 5-thio-dinitrobenzoic acid anion, which presents with a stable yellow color. GSH can be calculated by measuring the absorbance of the anions in solution at 412 nm.

Key features
• Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
• Broad range of kits—across key analytes and targets within the most important metabolic pathways
• Consistent—strict quality control criteria to ensure reproducible and reliable results

Specifications
Assay Range17.17–518.32 U/L
Assay Sensitivity17.17 U/L
For Use With (Equipment)Microplate Reader
Incubation Time10 min.
Interassay CV6.2
Intraassay CV2.4
Quantity96 tests
Sample Volume40 μL
Shipping ConditionDry Ice, 2–8°C
Target Kit NamedGlutathione Peroxidase (GSH-Px)
Total Assay Time40 min.
Detection MethodColorimetric
Sample TypeCell Culture Supernatant, Plasma, Serum, Tissue Homogenates, Whole Cells
Unit SizeEach
Contents & Storage
Store at 2–8°C.

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

0 results displayed, search above for a specific certificate

Safety Data Sheets

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection