γ-Glutamyl Transferase (GGT/γ-GT) Colorimetric Activity Kit
γ-Glutamyl Transferase (GGT/γ-GT) Colorimetric Activity Kit
Invitrogen™

γ-Glutamyl Transferase (GGT/γ-GT) Colorimetric Activity Kit

The γ-Glutamyl Transferase (GGT/γ-GT) Colorimetric Activity Kit is a highly reliable and sensitive assay designed for the quantitative measurement ofRead more
Have Questions?
Catalog NumberQuantity
EEA02996 tests
Catalog number EEA029
Price (USD)
696.00
Each
Add to cart
Quantity:
96 tests
Recurring order eligible. Learn more »
Price (USD)
696.00
Each
Add to cart
Ask our AI about this Product

The γ-Glutamyl Transferase (GGT/γ-GT) Colorimetric Activity Kit is a highly reliable and sensitive assay designed for the quantitative measurement of GGT in serum, plasma, and animal tissue samples. GGT is widely present in various organs of the human body, including the liver, kidney, and pancreas. GGT catalyzes the degradation of glutathione (GSH) and is involved in the regulation of glutathione levels in tissues, as well as the absorption and excretion of amino acids. Changes in GGT activity have been associated with various diseases, particularly those affecting the hepatobiliary system.

The assay utilizes the catalytic property of GGT to transfer a γ-glutamyl group from glutamyl p-nitroaniline to N-glycyl glycine, resulting in the production of p-nitroaniline. The p-nitroaniline has a characteristic absorption peak at 405 nm, allowing for the measurement of GGT activity.

Key features
• Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
• Broad range of kits—across key analytes and targets within the most important metabolic pathways
• Consistent—strict quality control criteria to ensure reproducible and reliable results

Specifications
Assay Range0.88–399.4 U/L
Assay Sensitivity0.88 U/L
For Use With (Equipment)Microplate Reader
Incubation Time6 min.
Interassay CV6.2
Intraassay CV4.2
Quantity96 tests
Sample Volume25 μL
Shipping ConditionDry Ice, 2–8°C
Target Kit Namedγ-Glutamyl Transferase (GGT/γ-GT)
Total Assay Time20 min.
Detection MethodColorimetric
Sample TypePlasma, Serum, Tissue Homogenates
Unit SizeEach
Contents & Storage
Store at 2–8°C.

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

0 results displayed, search above for a specific certificate

Other products to consider



Share catalog number, name or link