T4 DNA Ligase (5 U/μL)
T4 DNA Ligase (5 U/μL)
Thermo Scientific™

T4 DNA Ligase (5 U/μL)

Thermo Scientific T4 DNA Ligase catalyzes the formation of a phosphodiester bond between juxtaposed 5'-phosphate and 3'-hydroxyl termini in duplexRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
EL00111,000 units
EL00125 x 1,000 units
EL0014200 units
Catalog number EL0011
Price (USD)
252.65
Online Exclusive
301.00
Save 48.35 (16%)
Each
Add to cart
Quantity:
1,000 units
Request bulk or custom format
Price (USD)
252.65
Online Exclusive
301.00
Save 48.35 (16%)
Each
Add to cart
Thermo Scientific T4 DNA Ligase catalyzes the formation of a phosphodiester bond between juxtaposed 5'-phosphate and 3'-hydroxyl termini in duplex DNA or RNA. The enzyme repairs single-strand nicks in duplex DNA, RNA, or DNA/RNA hybrids. It also joins DNA fragments with either cohesive or blunt termini, but has no activity on single-stranded nucleic acids.

T4 DNA Ligase requires ATP as a cofactor.

Highlights

• Active in Themo Scientific restriction enzyme, PCR, and RT buffers (when supplemented with ATP)
• Fast—sticky-end ligation is completed in 10 minutes at room temperature
• Supplied with PEG solution for efficient blunt-end ligation

Applications

• Cloning of restriction enzyme generated DNA fragments
• Cloning of PCR products
• Joining of double-stranded oligonucleotide linkers or adaptors to DNA
• Site-directed mutagenesis
• Amplified fragment length polymorphism (AFLP)
• Ligase-mediated RNA detection (see Reference 3)
• Nick repair in duplex DNA, RNA or DNA/RNA hybrids
• Self-circularization of linear DNA.

Includes

• T4 DNA Ligase
• 10X T4 DNA Ligase Buffer
• 50% PEG Solution

Notes

• Binding of T4 DNA Ligase to DNA may result in a band shift in agarose gels. To avoid this, incubate samples with 6X DNA Loading Dye & SDS Solution at 70°C for 5 min or 65°C for 10 minutes and chill on ice prior to electrophoresis.
• The volume of the ligation reaction mixture should not exceed 10% of the competent cell volume in the transformation process.
• Prior to electro-transformation, remove T4 DNA Ligase from the ligation mixture using spin columns or chloroform extraction. The extracted DNA can be further precipitated with ethanol.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Compatible Buffer10X T4 DNA Ligase Buffer
Product TypeT4 DNA Ligase
Quantity1,000 units
Concentration5 U/μL
EnzymeDNA Ligase
Unit SizeEach

Frequently asked questions (FAQs)

What are white precipitates in the Thermo Scientific 10X T4 DNA Ligase Buffer that appear upon thawing?

The white precipitates are DTT in the buffer which may appear after freezing. To redissolve DTT, warm up the buffer to 37 degrees C by holding the vial between your palms, or incubating in a water bath for a few minutes, and mix well. The formation of DTT precipitates does not influence the quality of the buffer. The 10X T4 DNA Ligase Buffer is also available separately (Cat. No. B69).

What is the difference between Rapid DNA Ligation Kit and Thermo Scientific T4 DNA ligase with a standard buffer?

The speed of ligation is the main difference between the two. The Thermo Scientific Rapid DNA Ligation Kit enables fast sticky-end or blunt-end DNA ligation in only 5 minutes at room temperature. The kit contains T4 DNA ligase and a specially-formulated 5X rapid ligation buffer optimized for fast and efficient DNA ligation. Fast ligation efficiency is equal to that obtained with T4 DNA ligase in a standard 1 hour ligation.

Why do I see differences in the units of T4 DNA ligase from Thermo Scientific and other vendors?

Thermo Scientific T4 DNA ligase activity is expressed in Weiss units. One Weiss unit is equivalent to approximately 200 cohesive end units (CEU) which may be used by other vendors. One Weiss unit of the enzyme catalyzes the conversion of 1 nmol of [32PPi] into Norit-adsorbable form in 20 min at 37 degrees C. One CEU is defined as the amount of enzyme required to give 50% ligation of HindIII fragments of lambda DNA in 30 min at 16 degrees C.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link