T4 DNA Ligase (5 U/μL)
T4 DNA Ligase (5 U/μL)
Thermo Scientific™

T4 DNA Ligase (5 U/μL)

Thermo Scientific T4 DNA Ligase catalyzes the formation of a phosphodiester bond between juxtaposed 5'-phosphate and 3'-hydroxyl termini in duplexRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
EL0014200 units
EL00111,000 units
EL00125 x 1,000 units
Catalog number EL0014
Price (EUR)
46,65
Online Exclusive
49,65
Save 3,00 (6%)
Each
-
Add to cart
Quantity:
200 units
Request bulk or custom format
Price (EUR)
46,65
Online Exclusive
49,65
Save 3,00 (6%)
Each
Add to cart
Ask our AI about this Product
Thermo Scientific T4 DNA Ligase catalyzes the formation of a phosphodiester bond between juxtaposed 5'-phosphate and 3'-hydroxyl termini in duplex DNA or RNA. The enzyme repairs single-strand nicks in duplex DNA, RNA, or DNA/RNA hybrids. It also joins DNA fragments with either cohesive or blunt termini, but has no activity on single-stranded nucleic acids.

T4 DNA Ligase requires ATP as a cofactor.

Highlights

• Active in Themo Scientific restriction enzyme, PCR, and RT buffers (when supplemented with ATP)
• Fast—sticky-end ligation is completed in 10 minutes at room temperature
• Supplied with PEG solution for efficient blunt-end ligation

Applications

• Cloning of restriction enzyme generated DNA fragments
• Cloning of PCR products
• Joining of double-stranded oligonucleotide linkers or adaptors to DNA
• Site-directed mutagenesis
• Amplified fragment length polymorphism (AFLP)
• Ligase-mediated RNA detection (see Reference 3)
• Nick repair in duplex DNA, RNA or DNA/RNA hybrids
• Self-circularization of linear DNA.

Includes

• T4 DNA Ligase
• 10X T4 DNA Ligase Buffer
• 50% PEG Solution

Notes

• Binding of T4 DNA Ligase to DNA may result in a band shift in agarose gels. To avoid this, incubate samples with 6X DNA Loading Dye & SDS Solution at 70°C for 5 min or 65°C for 10 minutes and chill on ice prior to electrophoresis.
• The volume of the ligation reaction mixture should not exceed 10% of the competent cell volume in the transformation process.
• Prior to electro-transformation, remove T4 DNA Ligase from the ligation mixture using spin columns or chloroform extraction. The extracted DNA can be further precipitated with ethanol.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Compatible Buffer10X T4 DNA Ligase Buffer
Product TypeT4 DNA Ligase
Quantity200 units
Concentration5 U/μL
EnzymeDNA Ligase
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link