RNase H (5 U/μL)
Inquire about OEM or Commercial Supply version of this product here.
RNase H (5 U/μL)
Thermo Scientific™

RNase H (5 U/μL)

Thermo Scientific Ribonuclease H (RNase H) specifically degrades the RNA strand in RNA-DNA hybrids. It does not hydrolyze the phosphodiesterRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
EN0201100 units
EN0202500 units
Catalog number EN0201
Price (USD)/ Each
215.65
Online exclusive
256.00 
Save 40.35 (16%)
-
Add to cart
Quantity:
100 units
Request bulk or custom format
Price (USD)/ Each
215.65
Online exclusive
256.00 
Save 40.35 (16%)
Add to cart
RNase H (5 U/μL)
Catalog numberEN0201
Price (USD)/ Each
215.65
Online exclusive
256.00 
Save 40.35 (16%)
-
Add to cart
Thermo Scientific Ribonuclease H (RNase H) specifically degrades the RNA strand in RNA-DNA hybrids. It does not hydrolyze the phosphodiester bonds within single-stranded and double-stranded DNA and RNA.

Applications

• Removal of mRNA prior to synthesis of second strand cDNA
RT-PCR and qRT-PCR: removal of RNA after first strand cDNA synthesis
• Removal of the poly(A) sequences of mRNA after hybridization with oligo(dT)
• Site-specific cleavage of RNA
• Studies of in vitro polyadenylation reaction products
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration5 U/μL
EnzymeRNase
Compatible Buffer10X Reaction Buffer
Quantity100 units
Product TypeRNase H
Unit SizeEach