S1 Nuclease (100 U/μL)
S1 Nuclease (100 U/μL)
Thermo Scientific™

S1 Nuclease (100 U/μL)

Thermo Scientific S1 Nuclease degrades single-stranded nucleic acids, releasing 5'-phosphoryl mono- or oligonucleotides. It is five times more active onRead more
Promo ImagePromotion
Promo code:P5906365
Don't miss out: buy 3, only pay for 2
Promo ImagePromotion
Promo code:RPUZZ25
Stock up on essentials to piece your discovery together
Have Questions?
Catalog NumberQuantity
EN0321Promo Image10,000 units
Catalog number EN0321
Price (USD)
69.65
Online Exclusive
87.25
Save 17.60 (20%)
Each
In stock
Add to cart
Quantity:
10,000 units
Request bulk or custom format
Price (USD)
69.65
Online Exclusive
87.25
Save 17.60 (20%)
Each
Add to cart
Thermo Scientific S1 Nuclease degrades single-stranded nucleic acids, releasing 5'-phosphoryl mono- or oligonucleotides. It is five times more active on DNA than on RNA. S1 Nuclease also cleaves dsDNA at the single-stranded region caused by a nick, gap, mismatch or loop. S1 Nuclease exhibits 3'-phosphomonoesterase activity.

The enzyme is a glycoprotein with carbohydrate content of 18%.

Applications

• Removal of single-stranded overhangs of DNA fragments
• S1 transcript mapping
• Cleavage of hairpin loops
• Creation of unidirectional deletions in DNA fragments in conjunction with Exonuclease III

Note

S1 Nuclease can introduce breaks into double-stranded DNA, RNA and DNA/RNA hybrids at high enzyme and low salt concentrations.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Compatible BufferStorage Buffer, 5X Reaction Buffer
Product TypeS1 Nuclease
Quantity10,000 units
Concentration100 U/μL
EnzymeNuclease
Unit SizeEach

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link