Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

Uracil-DNA Glycosylase (1 U/μL)

Catalog number: EN0362
Have Questions?
Thermo Scientific™

Uracil-DNA Glycosylase (1 U/μL)

Catalog number: EN0362
Have Questions?
Catalog Number
EN0362
Unit Size
5 x 200 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
200 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
EN03625 x 200 units
-
EN0361200 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Uracil-DNA Glycosylase (UDG, UNG) catalyzes the hydrolysis of the N-glycosylic bond between uracil and sugar, leaving an apyrimidinic site in uracil-containing single or double-stranded DNA (see Figure 1 in Supporting Data). The enzyme shows no activity on RNA.

Highlights

Active in Thermo Scientific buffers for restriction enzymes and thermophilic polymerases

Applications

• Control of carry-over contamination in PCR
• Glycosylase mediated single nucleotide polymorphism detection (GMPD)
• Site-directed mutagenesis
• As a probe for protein-DNA interaction studies
• SNP genotyping
• Cloning of PCR products
• Generation of single strand overhangs of PCR products and cDNA

Note

The abasic sites formed in DNA by Uracil-DNA Glycosylase may be subsequently cleaved by heat, alkali-treatment or endonucleases that cleave specifically at abasic sites. UDG (UNG) is active in the presence or absence of divalent cations.

Use of this enzyme in certain applications may be covered by patents and may require a license.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
1 U/μL
Enzyme
Uracil-DNA Glycosylase
Compatible Buffer
10X Reaction Buffer
Quantity
5 x 200 U
Product Type
Uracil DNA Glycosylase
Unit Size
5 x 200 units

Contents & Storage

Uracil-DNA glycolase (1 U/μL) and 10X Reaction Buffer. Store at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text