RNase T1 (1000 U/μL)
Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™
RNase T1 (1000 U/μL)
Thermo Scientific RNase T1 is an endoribonuclease that specifically degrades single-stranded RNA at G residues. It cleaves the phosphodiester bondRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
EN0542500,000 U
EN0541100,000 U
2 Options
Catalog number EN0542
Price (USD)/ 500,000 units
297.65
Online exclusive
335.00 
Save 37.35 (11%)
-
Add to cart
Quantity:
500,000 U
Request bulk or custom format
Price (USD)/ 500,000 units
297.65
Online exclusive
335.00 
Save 37.35 (11%)
Add to cart
RNase T1 (1000 U/μL)
Catalog numberEN0542
Price (USD)/ 500,000 units
297.65
Online exclusive
335.00 
Save 37.35 (11%)
-
Add to cart
Thermo Scientific RNase T1 is an endoribonuclease that specifically degrades single-stranded RNA at G residues. It cleaves the phosphodiester bond between the 3'-guanylic residue and the 5'-OH residue of adjacent nucleotides with the formation of corresponding intermediate 2', 3'-cyclic phosphate. The reaction products are 3'-GMP and oligonucleotides with a terminal 3'-GMP.

RNase T1 does not require metal ions for activity.

Applications

• Removal of RNA from DNA preparations
• RNA sequencing
• Ribonuclease protection assays. Used in conjunction with RNase A
• Removal of RNA from recombinant protein preparations
• Determination of the level of RNA transcripts synthesized in vitro from DNA templates containing a 'G-less cassette'
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration1000 U/μL
EnzymeRNase
Quantity500,000 U
Product TypeRNase T1
Unit Size500,000 units