Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

RNase I (10 U/μL)

Catalog number: EN0602
Have Questions?
Thermo Scientific™

RNase I (10 U/μL)

Catalog number: EN0602
Have Questions?
Catalog Number
EN0602
Unit Size
5,000 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1,000 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
EN06025,000 units
-
EN06011,000 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Ribonuclease I (RNase I) is an endoribonuclease that preferentially hydrolyzes single-stranded RNA to nucleoside 3'-monophosphates via nucleoside 2', 3'-cyclic monophosphate intermediates.

The enzyme does not require any metal ions for activity.

Highlights

• Stable and active under a wide variety of reaction conditions
• Can be heat inactivated in 30 minutes at 100°C

Applications

• Removal of RNA from DNA solutions
• Removal of RNA from recombinant protein preparations
• Ribonuclease protection assays

Notes

Mammalian ribonuclease inhibitors have no effect on RNase I. RNase I binds to DNA, but does not degrade it. RNase I amino acid sequence is typical for the RNase T2 family. Non-ionic detergents (e.g., Triton X-100) do not inhibit RNase I, and may even slightly stimulate its activity and stabilize it against heat inactivation. Triton X-100 or BSA (at 0.1 mg/mL) may prevent RNase I from sticking to glass vessels when working with dilute solutions. Polyamines stimulate the activity of RNase I.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
10 U/μL
Enzyme
RNase
Quantity
5000 U
Product Type
RNase I

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text