Informational:Inquire about OEM or Commercial Supply version of this product here.
Informational:GMP-quality RNase inhibitor is available for RNA vaccine and therapeutic development. See details
Thermo Scientific™

RiboLock RNase Inhibitor (40 U/μL)

Catalog number: EO0382
Have Questions?
Thermo Scientific™

RiboLock RNase Inhibitor (40 U/μL)

Catalog number: EO0382
Have Questions?
Catalog Number
EO0382
Unit Size
4 x 2,500 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
2,500 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
24 x 2,500 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
EO03824 x 2,500 units
-
EO03812,500 units
-
EO038424 x 2,500 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific RiboLock RNase Inhibitor inhibits the activity of RNases A,B and C by binding them in a noncompetitive mode at a 1:1 ratio. It does not inhibit eukaryotic RNases T1, T2, U1, U2, CL3 as well as prokaryotic RNases I and H.

Highlights

Performs under a wide range of reaction conditions
Protects RNA from degradation at temperatures up to 55°C

Note

DTT provided in the Storage Buffer ensures stability during long term storage, but is not necessary for inhibitor activity. Recommended concentration 1 U/ μL of a reaction mixture.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
40 U/μL
Compatible Buffer
Storage Buffer
Quantity
4 x 2500 U
Product Type
RNase Inhibitor
Product Line
RiboLock™

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text