Proteinase K, recombinant, PCR grade
Proteinase K, recombinant, PCR grade
Thermo Scientific™

Proteinase K, recombinant, PCR grade

Thermo Scientific Proteinase K is a broad-range endolytic protease widely used for digestion of proteins in nucleic acid preparations. ItRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
EO04911 mL
EO04925 x 1.0 mL
Catalog number EO0491
Price (EUR)
40,65
Online Exclusive
46,33
Save 5,68 (12%)
Each
-
Add to cart
Quantity:
1 mL
Recurring order eligible. Learn more »
Request bulk or custom format
Price (EUR)
40,65
Online Exclusive
46,33
Save 5,68 (12%)
Each
Add to cart
Ask our AI about this Product
Thermo Scientific Proteinase K is a broad-range endolytic protease widely used for digestion of proteins in nucleic acid preparations. It degrades proteins even in the presence of detergents. Proteinase K cleaves peptide bonds at the carboxylic sides of aliphatic, aromatic, or hydrophobic amino acids. The Proteinase K is classified as a serine protease. The smallest peptide to be hydrolyzed by this enzyme is a tetrapeptide.

Highlights

• Ready-to-use solution
• Active in a wide range of reaction conditions

Applications

• Isolation of genomic DNA from mouse tail
• Isolation of genomic DNA from cultured cells
• Removal of DNases and RNases when isolating DNA and RNA from tissues or cell lines
• Determination of enzyme localization
• Improving cloning efficiency of PCR products

Note

• The recommended working concentration of Proteinase K is 0.05 to 1 mg/mL. The activity of the enzyme is stimulated by 0.2 to 1% SDS or by 1 to 4 M urea
• Ca2+ protects Proteinase K against autolysis, increases the thermal stability, and has a regulatory function for the substrate binding site of Proteinase K
• Stable over a wide pH range: 4.0 to 12.5, optimum pH 7.5 to 8.0
• Optimum activity at 50 to55°C
• Rapid denaturation of enzyme occurs at temperatures above 65°C
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeProteinase K
Quantity1 mL
Concentration>600 U/mL (∼20 mg/mL)
FormLiquid
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
Plate warping is causing problems with the robot in my automated processes. How can I solve this?
Are Thermo Scientific ABgene plastics sterile?
What 96-well plates do you recommend for -80 degrees C storage?
Are Thermo Scientific ABgene plastics endotoxin-free?
I would like to use the flat ultra-clear and domed cap strips for Thermo-Fast PCR Plate, 48-well, clear (Cat. No. AB0648). What do you recommend?
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Kunden, die diesen Artikel ansahen, interessierten sich auch für



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3269325Certificate of AnalysisJun 26, 2025EO0492
3263002Certificate of AnalysisJun 18, 2025EO0491
3255205Certificate of AnalysisJun 06, 2025EO0492
3250453Certificate of AnalysisMay 30, 2025EO0492
3247445Certificate of AnalysisMay 27, 2025EO0492
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

The recommended working concentration of Proteinase K is 0.05 to 1 mg/mL.

Andere Produkte



Share catalog number, name or link

1x1 image pixel for data collection