Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

T4 beta-glucosyltransferase

Catalog number: EO0831
Have Questions?
Thermo Scientific™

T4 beta-glucosyltransferase

Catalog number: EO0831
Have Questions?
Catalog Number
EO0831
Unit Size
500 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
EO0831500 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific T4 β-glucosyltransferase (T4 BGT) transfers the glucose moiety of uridine diphosphoglucose (UDP-glucose) to the 5-hydroxymethylcytosine (5-hmC) residues in double-stranded DNA generating ß-glucosyl-5 hydroxymethylcytosine.

Thermo Scientific T4 BGT is specifically formulated for fast reaction times without compromising the reaction efficiency. The enzyme completes 5-hmC glucosylation of 1 μg DNA at 37°C in 15 minutes.

Highlights

Specific—selectively transfers glucose to the hydroxymethyl moiety of 5-hmC
Fast—complete glucosylation of 1 μg DNA in just 15 min
Convenient—supplied with optimized buffer and UDP-glucose.

Applications

• Locus specific detection of 5-hmC
• Enrichment of 5-hmC containing DNA
• Labeling of 5-hmC residues using a radioactive UDP-glucose donor

Includes
• T4 β-glucosyltransferase, 5 U/μL
• 10X Epi buffer
• 10X UDP-glucose
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Enzyme
T4 β-glucosyltransferase
Compatible Buffer
10X Epi Buffer
Quantity
500 U
Product Type
T4 Beta Glucosyltransferase
Unit Size
500 units

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text