Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

DNA Polymerase I (10 U/μL)

Catalog number: EP0041
Have Questions?
Thermo Scientific™

DNA Polymerase I (10 U/μL)

Catalog number: EP0041
Have Questions?
Catalog Number
EP0041
Unit Size
500 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
2,500 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
EP0041500 units
-
EP00422,500 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific DNA Polymerase I, a template-dependent DNA polymerase, catalyzes 5'→3' synthesis of DNA. The enzyme also exhibits 3'→5' exonuclease (proofreading) activity, 5'→3' exonuclease activity, and ribonuclease H activity.

Highlights

• Incorporates modified nucleotides (e.g. biotin-, digoxigenin-, aminoallyl-, fluorescently-labeled nucleotides)
• Active in multiple buffers, including restriction enzyme, PCR, and RT buffers

Applications

• DNA labeling by nick-translation in conjunction with DNase
• Second-strand synthesis of cDNA in conjunction with RNase H
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Description
DNA Polymerase I
Polymerase
DNA Polymerase I
Quantity
500 U
Concentration
10 U/μL

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text