Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

T4 DNA Polymerase (5 U/μL)

Catalog number: EP0061
Have Questions?
Thermo Scientific™

T4 DNA Polymerase (5 U/μL)

Catalog number: EP0061
Have Questions?
Catalog Number
EP0061
Unit Size
100 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
EP0061100 units
-
EP0062500 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific T4 DNA Polymerase is a template-dependent DNA polymerase that catalyzes 5'-3' synthesis from primed single-stranded DNA. The enzyme has a 3'-5' exonuclease activity, but lacks 5'-3' exonuclease activity.

Highlights

Stronger 3'-5' exonuclease activity on single-stranded than on double-stranded DNA and greater (more than 200 times) than DNA polymerase I, E. coli,and Klenow fragment
• Active in Thermo Scientific restriction enzyme, PCR, RT and T4 DNA Ligase buffers

Applications

• Blunting of DNA ends: fill-in of 5'-overhangs or/and removal of 3'-overhangs(see​ References1, 2)
• Blunting of PCR products with 3'-dA overhangs
• Synthesis of labeled DNA probes by the replacement reaction(see​ Reference3)
• Oligonucleotide-directed site-specific mutagenesis(see​ Reference4)
• Ligation-independent cloning of PCR products
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Polymerase
T4 DNA Polymerase
Quantity
100 unit
Includes
1mL of 5X Reaction Buffer

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text