Terminal Deoxynucleotidyl Transferase (20 U/μL)
Terminal Deoxynucleotidyl Transferase (20 U/μL)
Thermo Scientific™

Terminal Deoxynucleotidyl Transferase (20 U/μL)

Thermo Scientific Terminal Deoxynucleotidyl Transferase (TdT) is a template-independent DNA polymerase that catalyzes the repetitive addition of deoxyribonucleotides to theRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
EP0161500 units
EP01622,500 units
Catalog number EP0161
Price (USD)
99.65
Online Exclusive
113.00
Save 13.35 (12%)
Each
-
Add to cart
Quantity:
500 units
Request bulk or custom format
Price (USD)
99.65
Online Exclusive
113.00
Save 13.35 (12%)
Each
Add to cart
Ask our AI about this Product
Thermo Scientific Terminal Deoxynucleotidyl Transferase (TdT) is a template-independent DNA polymerase that catalyzes the repetitive addition of deoxyribonucleotides to the 3'-OH of oligodeoxyribonucleotides and single-stranded and double-stranded DNA. TdT requires an oligonucleotide of at least three nucleotides to serve as a primer. With RNA as template TdT shows variable performance which strongly depends upon the tertiary structure of acceptor RNA 3'-end and the nature of nucleotide. Generally, it is lower than using DNA as a template.

Highlights

Incorporates modified nucleotides (e.g., fluorescein-, biotin-, aminoallyl-labeled nucleotides)

Applications

• Production of synthetic homo- and heteropolymers
• Homopolymeric tailing of linear duplex DNA with any type of 3'-OH terminus
• Oligodeoxyribonucleotide and DNA labeling
• 5'-RACE (Rapid Amplification of cDNA Ends)
In situ localization of apoptosis

Note

Due to the presence of CoCl2, the TdT Reaction Buffer is incompatible with downstream applications. It is necessary to remove CoCl2 from the reaction mixture by spin column or phenol/chloroform extraction and subsequent ethanol precipitation.
WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Compatible BufferStorage Buffer, 5X Reaction Buffer
Product TypeTdT
Quantity500 units
Concentration20 U/μL
EnzymeTerminal Deoxynucleotidyl Transferase
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

Is the Terminal Deoxynucleotidyl Transferase (TdT) within the Pierce Biotin 3' End DNA Labeling Kit (Cat. No. 89818) available separately?

Yes, but not the specific one from that same kit. We have the following comparable Terminal Deoxynucleotidyl Transferases (Cat. No. 10533065, 10533073 or EP0161, EP0162).

Find additional tips, troubleshooting help, and resources within our Nucleic Acid Gel Electrophoresis and Blotting Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link