Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

Taq DNA Polymerase, recombinant (5 U/μL)

Catalog number: EP0402
Have Questions?
Thermo Scientific™

Taq DNA Polymerase, recombinant (5 U/μL)

Catalog number: EP0402
Have Questions?
Catalog Number
EP0402
Unit Size
500 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 x 500 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
5 x 500 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
EP0402500 units
-
EP0401100 units
-
EP040610 x 500 units
-
EP04055 x 500 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Taq DNA Polymerase is a highly thermostable DNA polymerase from the thermophilic bacterium Thermus aquaticus. The enzyme catalyzes 5'→3' synthesis of DNA, has no detectable 3'→5' exonuclease (proofreading) activity and possesses low 5'→3' exonuclease activity. In addition, Taq DNA Polymerase exhibits deoxynucleotidyl transferase activity, which frequently results in the addition of extra adenines at the 3'-end of PCR products. Recombinant Taq DNA Polymerase is the ideal tool for standard PCR of templates 5 kb or shorter.

Highlights

Thermostable—half life is more than 40 min at 95°C
Generates PCR products with 3'-dA overhangs
Supplied with two buffers—10X Taq Buffer with KCl and 10X Taq Buffer with (NH4)2SO4. The latter allows for PCR at wide range of magnesium concentrations and decreases unspecific priming
Incorporates modified nucleotides (e.g., biotin-, digoxigenin-, fluorescently-labeled nucleotides)

Applications

• Routine PCR amplification of DNA fragments up to 5 kb
• High throughput PCR
• DNA labeling

Note

• The error rate of Taq DNA Polymerase in PCR is 2.2 x 10-5 errors per nt per cycle. Accordingly, the accuracy of PCR is 4.5 x 104. Accuracy is an inverse of the error rate and shows an average number of correct nucleotides incorporated before an error occurs.
• The 10X Taq Buffer without Detergent is recommended for microarray experiments.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

GC-Rich PCR Performance
Low
Polymerase
Taq DNA Polymerase
Reaction Speed
Standard
Product Type
Taq DNA Polymerase (recombinant) (5 U/μL)
Quantity
500 Units
Fidelity (vs. Taq)
1 X
Hot Start
No
Overhang
3'-A
Reaction Format
Standalone

Contents & Storage

Contains:
• 100 μL Taq DNA Polymerase (5 U/μL)
• 2 x 1.25 mL 10X Taq Buffer with KCl
• 2 x 1.25 mL 10X Taq Buffer with (NH4)2SO4
• 2 x 1.25 mL 25 mM MgCl2

Store at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text