Taq DNA Polymerase, recombinant (1 U/μL)
<i>Taq</i> DNA Polymerase, recombinant (1 U/&mu;L)
Thermo Scientific™

Taq DNA Polymerase, recombinant (1 U/μL)

Thermo Scientific Taq DNA Polymerase is a highly thermostable DNA polymerase from the thermophilic bacterium Thermus aquaticus.
Have Questions?
Catalog NumberQuantity
EP0404500 Units
Catalog number EP0404
Price (USD)
177.00
Each
-
Add to cart
Quantity:
500 Units
Request bulk or custom format
Price (USD)
177.00
Each
Add to cart
Ask our AI about this Product
Thermo Scientific Taq DNA Polymerase is a highly thermostable DNA polymerase from the thermophilic bacterium Thermus aquaticus. The enzyme catalyzes 5' to 3' synthesis of DNA, has no detectable 3' to 5' exonuclease (proofreading) activity, and possesses low 5' to 3' exonuclease activity. In addition, Taq DNA Polymerase exhibits deoxynucleotidyl transferase activity, which frequently results in the addition of extra adenines at the 3'-end of PCR products. Recombinant Taq DNA Polymerase is the ideal tool for standard PCR of templates 5 kb or shorter.

Taq DNA Polymerase is supplied with two buffers: 10X Taq Buffer with KCl and 10X Taq Buffer with (NH4)2SO4. The latter allows for PCR at wide range of magnesium concentrations and decreases unspecific priming.

Features

  • High thermostability with half-life greater than 40 min at 95°C
  • Generates PCR products with 3'-dA overhangs
  • Incorporates modified nucleotides (e.g., biotin-, digoxigenin-, fluorescent-labeled nucleotides)

Applications

  • Routine PCR amplification of DNA fragments up to 5 kb
  • High throughput PCR
  • DNA labeling

Notes

  • The error rate of Taq DNA Polymerase in PCR is 2.2 x 10-5 errors per nt per cycle. Accordingly, the accuracy of PCR is 4.5 x 104. Accuracy is an inverse of the error rate and shows an average number of correct nucleotides incorporated before an error occurs.
  • 10X Taq Buffer without Detergent is recommended for microarray experiments.
For Research Use Only. Not for use in diagnostic procedures.
In certain countries use of this product is covered by patents. Purchase of product in these countries includes non-transferable, limited license for using only this amount of product for the purchaser's own internal research.
Specifications
Concentration1 U/μL
Fidelity (vs. Taq)1X
Hot StartNo
Overhang3'-A
PolymeraseTaq DNA Polymerase
Product TypeTaq DNA Polymerase (recombinant)
Quantity500 Units
Reaction FormatSeparate Components
For Use With (Application)Standard PCR
GC-Rich PCR PerformanceLow
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Taq DNA Polymerase (1 U/μL), 500 μL
• 10X Taq Buffer with KCl, 2 x 1.25 mL
• 10X Taq Buffer with (NH4)2SO4, 2 x 1.25 mL
• 25 mM MgCl2, 2 x 1.25 mL

Store at -20°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link