Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

Klenow Fragment, exo– (5 U/μL)

Catalog number: EP0421
Have Questions?
Thermo Scientific™

Klenow Fragment, exo– (5 U/μL)

Catalog number: EP0421
Have Questions?
Catalog Number
EP0421
Unit Size
300 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1,500 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
EP0421300 units
-
EP04221,500 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Klenow Fragment, exo-, is the large fragment of DNA polymerase I . It exhibits 5'→3' polymerase activity, but lacks the 3'→5' and 5'→3' exonuclease activities of DNA Polymerase I. The 3'→5' exonuclease activity of the enzyme is eliminated by mutations in the 3'→5'-exonuclease active site.

Highlights

• Lacks 3'→5' exonuclease activity
• Incorporates modified nucleotides (e.g., Cy3-, Cy5-, fluorescein-, rhodamine-, aminoallyl-, biotin-labeled nucleotides)
• Active in restriction enzyme, PCR, and RT buffers

Applications
• Random-primed DNA labeling
• Labeling by fill-in 5'-overhangs of dsDNA
• Strand displacement amplification (SDA)
• DNA sequencing by the Sanger method

Note

Klenow Fragment, exo- is not recommended for DNA blunting reactions prior to DNA ligation since it frequently adds one or more extra nucleotides to the 3'-terminus of blunt-end DNA substrates in a non-template directed fashion.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
5 U/μL
Polymerase
DNA Polymerase I
Compatible Buffer
Restriction Enzyme, PCR, RT Buffer
Quantity
300 U
Product Type
Klenow Fragment

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text