Informational:Inquire about OEM or Commercial Supply version of this product here.
Informational:Informational:Maxima H Minus cDNA Synthesis Master Mix now available. Get a sample »
Thermo Scientific™

Maxima Reverse Transcriptase (200 U/μL)

Catalog number: EP0743
Have Questions?
Thermo Scientific™

Maxima Reverse Transcriptase (200 U/μL)

Catalog number: EP0743
Have Questions?
Catalog Number
EP0743
Unit Size
4 x 10,000 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
2,000 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10,000 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
EP07434 x 10,000 units
-
EP07412,000 units
-
EP074210,000 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Thermo Scientific Maxima Reverse Transcriptase (RT) was developed through in vitro evolution of M-MuLV RT. The enzyme possesses an RNA-dependent and DNA-dependent polymerase activity as well as RNase H activity. The engineered enzyme features dramatically improved thermostability and robustness, and increased synthesis rates compared to wild type M-MuLV RT.

Maxima Reverse Transcriptase is capable of reproducible cDNA synthesis from a wide range of input total RNA amounts (from 1 pg to 5 μg) at elevated temperatures (50–65°C), which makes this enzyme an ideal tool for two step RT-qPCR (see supporting data).

Due to its high thermostability, Maxima enzyme maintains full activity during the entire reverse transcription reaction, generates the highest yields of cDNA, and is able to synthesize even very long RNA transcripts up to 20 kb. The reaction temperature can be increased up to 65°C for efficient transcription of RNA regions with a high secondary structure or to improve specificity using gene specific primers. Due to its increased synthesis rate, the reverse transcriptase reaction can be completed in 15 to 30 minutes

Features of Maxima Reverse Transcriptase include:
• High yields of full-length cDNA up to 20 kb
• Active up to 65°C
• Thermostable—90% active after incubation at 50°C for 60 minutes
• High sensitivity allows reproducible cDNA synthesis from a wide range of total RNA quantities (1 pg to 5 μg)
• Efficient formula for complete cDNA synthesis in 15 to 30 minutes
• Incorporates modified nucleotides

Applications
• Two step RT-PCR
• Two step RT-qPCR
• First strand cDNA synthesis
• Construction of full length cDNA libraries
• DNA labeling
• Primer extension

Also available
Maxima Reverse Transcriptase
Maxima First Strand cDNA Synthesis Kit for RT-qPCR
Maxima First Strand cDNA Synthesis Kit for RT-qPCR, with dsDNase

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Stand-alone Enzyme
GC-Rich PCR Performance
High
Reaction Speed
30 min.
Technique
Reverse Transcription
Optimal Reaction Temperature
50°C to 55°C
Quantity
4 x 10,000 Units
Reverse Transcriptase
Maxima
Ribonuclease H Activity
Yes
Shipping Condition
Dry Ice
Concentration
200 U/μL
Final Product Type
First-Strand cDNA
Reaction Format
Separate components
Reagent Type
Reverse Transcription
Size (Final Product)
Up to 20 kb
Starting Material
RNA

Contents & Storage

• Maxima Reverse Transcriptase, 4 x 10,000 units (200 U/μL)
• 5X RT Buffer

Store at –20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text