1x1 image pixel for data collection
EcoRI (10 U/μL)
EcoRI (10 U/μL)
Thermo Scientific™

EcoRI (10 U/μL)

5' G ↓A A T T C 3' 3' C T T A A ↑G 5' Thermo Scientific EcoRI restrictionRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
ER02715,000 units
ER02725 x 5,000 units
ER027510,000 units
Catalog number ER0271
Price (USD)
52.50
Each
Add to cart
Quantity:
5,000 units
Request bulk or custom format
Price (USD)
52.50
Each
Add to cart
5'  G ↓A  A  T  T  C   3' 
3'  C  T  T  A  A ↑G   5' 

Thermo Scientific EcoRI restriction enzyme recognizes G^AATTC sites and cuts best at 37°C in its own unique buffer. See Reaction Conditions for Restriction Enzymes for a table of enzyme activity, conditions for double digestion, and heat inactivation for this and other restriction enzymes. Note: Also available as a FastDigest enzyme for rapid DNA digestion.

Thermo Scientific conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that sent in DNA preparations.

Features

• Superior quality—stringent quality control and industry leading manufacturing process
• Convenient color-coded Five Buffer System
• Includes universal Tango buffer for double-digestions
• BSA premixed in reaction buffers
• Wide selection of restriction endonuclease specificities

Applications

• Molecular cloning
• Restriction site mapping
• Genotyping
• Southern blotting
• Restriction fragment length polymorphism (RFLP)
• SNP

Note: For methylation sensitivity, refer to product specifications.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Compatible BufferUnique Buffer (10X Buffer EcoRI )
Product TypeRestriction Enzyme
Quantity5,000 units
Concentration10 U/μL
EnzymeEcoR I
Methylation SensitivityCpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
Optimal Reaction Temperature37°C
Research CategoryTraditional Cloning
Sensitive to Heat InactivationYes
Type IIS RENo
Unit SizeEach

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection