Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

Eco57I (AcuI) (5 U/μL)

Catalog number: ER0341
Have Questions?
Thermo Scientific™

Eco57I (AcuI) (5 U/μL)

Catalog number: ER0341
Have Questions?
Catalog Number
ER0341
Unit Size
200 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1,000 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
ER0341200 units
-
ER03421,000 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
5'  C  T  G  A  A  G  N16 3'
3'  G  A  C  T  T  C  N14 5'

Thermo Scientific Eco57I (AcuI) restriction enzyme recognizes CTGAAG(16/14)^ sites and cuts best at 37°C in G (+SAM) buffer. See Reaction Conditions for Restriction Enzymes for a table of enzyme activity, conditions for double digestion, and heat inactivation for this and other restriction enzymes. Note: Also available as a FastDigest enzyme for rapid DNA digestion. Isoschizomers: AcuI.

Thermo Scientific conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that sent in DNA preparations.

Features

• Superior quality—stringent quality control and industry leading manufacturing process
• Convenient color-coded Five Buffer System
• Includes universal Tango buffer for double-digestions
• BSA premixed in reaction buffers
• Wide selection of restriction endonuclease specificities

Applications

• Molecular cloning
• Restriction site mapping
• Genotyping
• Southern blotting
• Restriction fragment length polymorphism (RFLP)
• SNP

Note: Eco57I requires only Mg2+ for its activity, but is stimulated by S-adenosylmethionine. 0.01 mM S-adenosylmethionine gives a 100-fold increase in Eco57I activity. Still, complete cleavage of some substrates with Eco57I is difficult to achieve Eco57I concentration is determined by the maximum cleavage level achieved when no change in the fragmentation pattern is observed with further enzyme increase. Low salt, high glycerol (> 5%) concentrations, pH > 8.0 or a large excess of enzyme may result in star activity. Eco57I may remain associated with the cleaved DNA. This may cause DNA band shifting during electrophoresis. To avoid atypical DNA band patterns, use the 6X DNA Loading Dye & SDS Solution for sample preparation or heat the digested DNA in the presence of SDS prior to electrophoresis.For methylation sensitivity, refer to product specifications.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
5 U/μL
Methylation Sensitivity
Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
Enzyme
Eco57I (AcuI)
Compatible Buffer
10X Buffer G
Sensitive to Heat Inactivation
Yes
Optimal Reaction Temperature
37°C
Type IIS RE
Yes
Quantity
200 U
Product Type
Restriction Enzyme
Research Category
Scarless Cloning

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text