GsuI (BpmI) (5 U/μL)
Inquire about OEM or Commercial Supply version of this product here.
GsuI (BpmI) (5 U/μL)
Thermo Scientific™

GsuI (BpmI) (5 U/μL)

5' C T G G A G N16↓ 3'3' G A C C T C N14↑ 5'Thermo Scientific GsuI (BpmI)Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
ER0462500 U
ER0461100 U
2 Options
Catalog number ER0462
Price (USD)/ 500 units
355.00
-
Add to cart
Quantity:
500 U
Request bulk or custom format
Price (USD)/ 500 units
355.00
Add to cart
GsuI (BpmI) (5 U/μL)
Catalog numberER0462
Price (USD)/ 500 units
355.00
-
Add to cart
5'  C  T  G  G  A  G  N16 3'
3'  G  A  C  C  T  C  N14 5'

Thermo Scientific GsuI (BpmI) restriction enzyme recognizes CTGGAG(16/14)^ sites and cuts best at 30°C in B buffer. See Reaction Conditions for Restriction Enzymes for a table of enzyme activity, conditions for double digestion, and heat inactivation for this and other restriction enzymes. Note: Also available as a FastDigest enzyme for rapid DNA digestion. Isoschizomers: BpmI.

Thermo Scientific conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

Features:

• Superior quality—stringent quality control and industry leading manufacturing process
• Convenient color-coded Five Buffer System
• Includes universal Tango buffer for double-digestions
• BSA premixed in reaction buffers
• Wide selection of restriction endonuclease specificities

Note:
Incubation at 37°C results in 70% activity.
GsuI requires only Mg2+ for its activity, but is stimulated by S-adenosylmethionine. 0.01 mM S-adenosylmethionine gives a 2-fold increase in activity.
GsuI may remain associated with the cleaved DNA. This may cause DNA band shifting during electrophoresis. To avoid atypical DNA band patterns, use the 6X DNA Loading Dye & SDS Solution for sample preparation or heat the digested DNA in the presence of SDS prior to electrophoresis.
GsuI is blocked by overlapping dcm methylation. To avoid dcm methylation, use a dam-, dcm- strain, such as GM2163.
For methylation sensitivity, refer to product specifications.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration5 U/μL
Methylation SensitivityDcm Methylation-Sensitive, Not Dam Methylation-Sensitive, Not CpG Methylation-Sensitive
EnzymeGsuI (BpmI)
Compatible Buffer10X Buffer B
Sensitive to Heat InactivationYes
Optimal Reaction Temperature30°C
Type IIS REYes
Quantity500 U
Product TypeRestriction Enzyme
Research CategoryScarless Cloning
Unit Size500 units