Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

SgeI (3 U/μL)

Catalog number: ER2211
Have Questions?
Thermo Scientific™

SgeI (3 U/μL)

Catalog number: ER2211
Have Questions?
Catalog Number
ER2211
Unit Size
250 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
ER2211250 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
5'  Cm5N  N  G  N9 3'
3'  G  N  N  C  N13 5'
SgeI cleaves DNA targets containing 5-methylcytosine on one or both DNA strands

Thermo Scientific SgeI restriction enzyme recognizes m5CNNG(9/13)^ sites and cuts best at 37°C in its own unique buffer. See Reaction Conditions for Restriction Enzymes for a table of enzyme activity, conditions for double digestion, and heat inactivation for this and other restriction enzymes.

Thermo Scientific conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

Features

• Superior quality—stringent quality control and industry leading manufacturing process
• Convenient color-coded Five Buffer System
• Includes universal Tango buffer for double-digestions
• BSA premixed in reaction buffers
• Wide selection of restriction endonuclease specificities

Applications

• Molecular cloning
• Restriction site mapping
• Genotyping
• Southern blotting
• Restriction fragment length polymorphism (RFLP)
• SNP

Note: DNA methylated by Dcm or CpG methyltransferases will be a substrate for SgeI. Greater than 3-fold overdigestion with SgeI may result in incomplete cleavage. At least two copies of SgeI recognition sequence are required for an efficient cleavage. Amount of the enzyme required for complete digestion of methylated DNA depends on the number of SgeI recognition sites. DNA cleavage products generated by target site cleavage facilitate the nonspecific cleavage by SgeI. Therefore, optimization of the enzyme amount is recommended for DNA cleavage. pBR322 DNA isolated from E. coli dcm+ strain (#SD0041) can be used as a DNA cleavage efficiency control. SgeI cleaves all six dcm methylated targets on pBR322 DNA. For methylation sensitivity, refer to product specifications.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
3 U/μL
Methylation Sensitivity
Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
Enzyme
SgeI
Compatible Buffer
Unique Buffer (10X Buffer SgeI)
Sensitive to Heat Inactivation
Yes
Optimal Reaction Temperature
37°C
Type IIS RE
No
Product Type
Restriction Enzyme
Research Category
Traditional Cloning

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text