Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Connect your nano or capillary column to your Thermo Scientific™ mass spectrometers (MS) more easily using Thermo Scientific™ EASY-Spray™ Nano & Capillary Emitters. The nano and capillary emitters act as a column-independent sprayer, allowing the introduction of low flow from nano and capillary columns without troublesome handling of traditional spray needles. There are two versions of emitters available: with and without integrated transfer lines. These sprayers are ideal for protein and peptide separation using nano and capillary HPLC.
Column Format | Capillary EASY-Spray Emitter |
Diameter (Metric) | Emitter: 15 μm I.D. |
Includes | Without Transfer Line |
Product Line | Easy Spray |
Column Type | Emitter |
Type | Capillary EASY-Spray Emitter, Bullet Type without transfer line |
Unit Size | Pack of 2 |
Column Format | EASY-Spray Emitter |
Diameter (Metric) | 0.02 mm |
Includes | Transfer line: 20 μm I.D. x 50 cm Length |
Length (Metric) | 500 mm |
Product Line | Easy Spray |
Packing Material | Unpacked |
Column Type | Emitter |
Type | Nano EASY-Spray Emitter with transfer line |
Unit Size | Each |
Column Format | EASY-Spray Emitter |
Diameter (Metric) | 0.075 mm |
Includes | Transfer line: 75 μm I.D. x 50 cm Length |
Length (Metric) | 500 mm |
Product Line | Easy Spray |
Packing Material | Unpacked |
Column Type | Emitter |
Type | Capillary EASY-Spray Emitter with transfer line |
Unit Size | Each |
Column Format | Nano EASY-Spray Emitter |
Diameter (Metric) | Emitter: 10 μm I.D. |
Includes | Without Transfer Line |
Product Line | Easy Spray |
Column Type | Emitter |
Type | Nano EASY-Spray Emitter, Bullet Type without transfer line |
Unit Size | Pack of 2 |
Catalog Number | Specifications | Unit Size | Diameter (Metric) | Type | Price (USD) | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
ES994 | Pack of 2 | Emitter: 15 μm I.D. | Capillary EASY-Spray Emitter, Bullet Type without transfer line | 818.85 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
ES991 | Each | 0.02 mm | Nano EASY-Spray Emitter with transfer line | 753.71 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
ES992 | Each | 0.075 mm | Capillary EASY-Spray Emitter with transfer line | 603.67 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
ES993 | Pack of 2 | Emitter: 10 μm I.D. | Nano EASY-Spray Emitter, Bullet Type without transfer line | Request A Quote | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
For sensitive protein and peptide separation using nano and capillary HPLC
These emitters offer easy connection to Thermo Scientific™ MAbPac™ and Accucore™ nano and capillary columns for enhanced separation and MS detection of proteins, peptides and mnoclonal antibodies (mAbs).