Search Thermo Fisher Scientific
Catalog Number | Type | Voltage |
---|---|---|
F01318 | Microplate Delidder, Seven Position Portrait | 115/230 V NA |
F01103 | Microplate Delidder, Single Nest | 115/230 V |
F01287 | Microplate Delidder, Seven Position Landscape | 115/230 V NA |
F01288 | Microplate Delidder, Seven Position Landscape | 115/230 V EU |
F01319 | Microplate Delidder, Seven Position Portrait | 115/230 V EU |
Automate the removal, storage, and replacement of lids on microplates using Thermo Scientific™ Microplate Delidder. Using a Microplate Delidder instead of a robot's microplate fingers to remove or replace a lid provides more reliable lid placement and frees the robot to perform other operations. As part of a Thermo Scientific Polara™ automated lab system, delidders enable the system to delid and relid any microplate being processed.