Thermo Scientific™

DynaChrom™ Single-Use Chromatography System

Catalog number: F100-3500-000
Have Questions?
Thermo Scientific™

DynaChrom™ Single-Use Chromatography System

Catalog number: F100-3500-000
Have Questions?
Catalog Number
F100-3500-000
Unit Size
Each
Price (USD)
Catalog NumberUnit SizePrice (USD)
F100-3500-000EachRequest A Quote
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific DynaChrom Chromatography System brings innovation, scalability, and robust automation to single-use downstream bioprocessing. It offers modular, single-use fluid transfer assemblies, a configurable system design with expandable features, and well-developed automation software built on an industry standard platform, enabling performance, flexibility, and speed in bioprocessing.

The system can be tailored for a specific process and flow rate(s) through the selection of low pulsation pumps, offered in three pump combinations, matched fluid transfer assembly sizes, and single-use valve block flow paths aligned with the fluid transfer assemblies. The novel single-use valve manifold design removes limitations caused by pinch valves or clam shell/liner designs, provides error-free fluid transfer assembly insertion, and enables up/down column flow, ensuring safe operation while providing a quicker turn-around time in process suites. The DynaChrom Chromatography System is designed to be column agnostic and can work with any end-user-selected column.

Packaged with our TruChrom software, the DynaChrom Chromatography System provides the ability to leverage existing site DeltaV network infrastructure and allows ease of tech transfer, data integration, and data storage. The system enables meeting of regulatory requirements for use in cGMP and CFR 21 Part 11 compliant processes. The use of robust components helps ensure reliable and consistent results. Modular design and simplified options help ensure scalability from process development to commercial production.

Features of the DynaChrom Chromatography System include:
• Unique manifold design
    --Easy and error-free fluid transfer assembly insertion
    --No-pinch-valve design
    --Enables up/down column flow
• Wide operational flow range
    --Flowrates from 1 L/hr to 1,980 L/hr depending on pump(s) selected
    --Fluid transfer assemblies in 1/4-, 3/8-, 1/2-, and 3/4-inch sizes
    --Low pulsation pumps
• Zero void volume/ring flush
    --Previous phase liquids are instantly flushed at the start of the next phase
    --Reduced buffer use
    --Improved peak resolution and product yield
• Built on the Emerson DeltaV automation platform
    --Ease of tech transfer and data integration
    --Ability to leverage existing DeltaV network infrastructure
    --Capable of running processes in batch mode
• Chromatography steps and modes of operation
    --Affinity, Anion Exchange (AEX), Cation Exchange (CEX), or Reversed Phased Chromatography (RPC)
    --Can be configured for bind/elute or flow-through modes of operation
    --Can support in-line dilution

Applications of the DynaChrom Chromatography System include:
• Monoclonal antibody and viral vector production
WARNING: Cancer – www.P65Warnings.ca.gov

Specifications

Max. Pressure
4 bar
Type
Single-Use Chromatography System
Flow Rate
1 to 1980 L/hr.
Pressure Range
0 to 4 bar
Weight (English)
1,200 lb.
Weight (Metric)
544.3 kg

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text