Invitrogen™

Fluo-4 Direct™ Calcium Assay Kit

Catalog number: F10471
Have Questions?
Invitrogen™

Fluo-4 Direct™ Calcium Assay Kit

Catalog number: F10471
Have Questions?
Catalog Number
F10471
Unit Size
10 x 10 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 x 100 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 x 1 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
F1047110 x 10 mL
-
F104721 x 100 mL
-
F104731 x 1 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Fluo-4 Direct™ Calcium Assay Kit was formulated to provide a homogeneous fluorescent calcium assay that:
1. Easily loads into cells
2. Achieves a large assay window and
3. Suppresses background fluorescence generated from the calcium indicator in complete media with little-to-no impact on the specific cellular fluorescence

This advanced formulation allows the assay to be run in a simple “addition only” format in the presence of serum-containing media. Fluo-4 Direct™ is the third addition to the Fluo-4 family of calcium detection reagents. Fluo-4 AM and Fluo-4 NW both require media removal before assay detection, but Fluo-4 NW adds convenience by including the PowerLoad™ reagent for ease in cell loading. The Fluo-4 Direct™ Calcium Assay Kit is similar to Fluo-4 NW in that it is formulated with PowerLoad™ for ease in cell loading, but unique in that it can be used in the presence of complete culture media and will efficiently suppress background fluorescence without sacrificing the specific cellular fluorescence generated in the assay.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color
Green
Detection Method
Fluorescent
For Use With (Application)
Calcium Assay
For Use With (Equipment)
Confocal Microscope, Fluorescence Microscope, High Content Analysis Instrument, HTS Reader, Microplate Reader, Fluorescent Imager
Product Type
Cell-based calcium flux
Dye Type
Fluorescent Dye-Based
Emission
Visible
Form
Powder
Modification
Chemical
Product Line
Molecular Probes™
Quantity
100 ml
Shipping Condition
Room Temperature
Sub Cellular Localization
Nucleus, Organelles, Cytoplasm

Contents & Storage

  • 10 × 10 ml Fluo-4 Direct™ Calcium Assay Reagent (Component A)
  • 2 × 77 mg Probenecid (Component B)
  • 200 ml Fluo-4 Direct™ Calcium Assay Buffer (Component C)
  • Store at ≤-20°C. Dessicate and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text