1x1 image pixel for data collection
Fluo-4 Calcium Imaging Kit
Fluo-4 Calcium Imaging Kit
Invitrogen™

Fluo-4 Calcium Imaging Kit

The Fluo-4 Calcium Imaging Kit has been designed for the specific detection of calcium flux by imaging applications. Upon bindingRead more
Have Questions?
Catalog NumberQuantity
F104891 kit
Catalog number F10489
Price (USD)
778.00
Each
In stock
Add to cart
Quantity:
1 kit
Price (USD)
778.00
Each
Add to cart
The Fluo-4 Calcium Imaging Kit has been designed for the specific detection of calcium flux by imaging applications. Upon binding of calcium, a large increase (>100-fold) in fluorescence emission can be detected. To minimize time to experimental results, this kit contains all of the reagents necessary for the detection of calcium flux from live cells.

Learn more about ion indicators including calcium, potassium, pH, and membrane potential indicators ›

Features of the Fluo-4 Calcium Imaging Kit include:

• Emission from the calcium-bound Fluo-4 dye (ex/em 494/506 nm) can be detected with standard FITC filters
• Upon binding calcium, fluorescence intensity increases >100-fold
• Kd for Ca2+ in buffer: ∼335 nM
• Contains all necessary reagents, reducing time to experimental results
• Exhibits fluorescence increase upon binding calcium with little shift in wavelength

All Necessary Reagents Included
The Fluo-4 Calcium Imaging Kit contains all of the necessary components for the detection of calcium flux by imaging applications. In addition to Fluo-4 dye, the kit contains PowerLoad™ Concentrate, a 10X Neuro Background Suppressor solution, and probenecid.

Labeled calcium indicators, such as Fluo-4 dye, are molecules that exhibit an increase in fluorescence upon binding calcium. Fluo-4 is an analog of Fluo-3, with two key chemical differences that result in an increased fluorescence excitation at 488 nm and consequently higher fluorescence signal levels (>100-fold increase in fluorescence intensity upon binding calcium). For ease of use, the Fluo-4 dye is supplied in a ready-to-use solution.

For easy cell loading, the Fluo-4 Calcium Imaging Kit includes PowerLoad™ Concentrate. Due to the unique nature of the PowerLoad™ solution, it can be used in the prescence of complete culture media, thus reducing the negative effects of replacing media or loading in serum-free media.

Baseline autofluorescence caused by components within growth media can be greatly reduced by the addition of the included Neuro Background Suppressor. This solution has been specifically formulated for use with neuronal cells and will not cause osmotic shock. Additionally, it has been used successfully with many different cell types to efficiently suppress background fluorescence without sacrificing the specific cellular fluorescence generated in the assay.

A proprietary, water-soluble probenecid (which is commonly used to inhibit cellular transport and thus reduces the baseline signal) is also supplied with the Fluo-4 Calcium Imaging Kit.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Dye TypeCalcium Indicator
Quantity1 kit
For Use With (Application)Calcium Assay
Product LineMolecular Probes
Product TypeCalcium Imaging Kit
Unit SizeEach
Contents & Storage
Contains:
  • 50 μL of Fluo-4 AM Dye in DMSO
  • 500 μL of PowerLoad™ Concentrate
  • 5 mL of Neuro Background Suppressor
  • 1 vial of Probenecid
  • Store at 2°C to 8°C
  • Do not freeze
  • Documents & Downloads

    Certificates

    Lot #Certificate TypeDateCatalog Number(s)
    2946750Certificate of AnalysisJul 11, 2024F10489
    2820938Certificate of AnalysisDec 14, 2023F10489
    2550808Certificate of AnalysisFeb 15, 2023F10489
    2456919Certificate of AnalysisFeb 16, 2022F10489
    2365683Certificate of AnalysisAug 09, 2021F10489
    5 results displayed, search above for a specific certificate

    Safety Data Sheets

    Product Information

    Other products to consider



    Share catalog number, name or link