Fura-2, AM is a high affinity, intracellular calcium indicator that is ratiometric and UV light—excitable. This acetoxymethyl (AM) ester form is useful for noninvasive intracellular loading and is also available in 1 mg amounts (F-1201) and in a DMSO solution (F-1225). Cell-impermeant pentapotassium (F-1200) and pentasodium (F-6799) salt forms of the dye are available.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection Method
Fluorescent
For Use With (Application)
Cell Viability, Proliferation and Cellular Imaging
For Use With (Equipment)
Fluorescence Microscope
Product Type
Calcium Indicator
Dye Type
Fluorescent Dye-Based
Quantity
1mg
Shipping Condition
Room Temperature
Unit Size
1 mg
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.