Invitrogen™

Fluo-3, AM, Calcium Indicator

Catalog number: F1242
Have Questions?
Invitrogen™

Fluo-3, AM, Calcium Indicator

Catalog number: F1242
Have Questions?
Catalog Number
F1242
Unit Size
20 x 50 µg
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
F124220 x 50 µg
-
F12411 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Labeled calcium indicators are molecules that exhibit an increase in fluorescence upon binding Ca2+. Fluo-3 has been used to image the spatial dynamics of Ca2+ signaling, in flow cytometry experiments involving photoactivation of caged chelators, second messengers, and neurotransmitters, and for cell-based pharmacological screening. Fluo-4 is an analog of fluo-3 with the two chlorine substituents replaced by fluorines, which results in increased fluorescence excitation at 488 nm and consequently higher fluorescence signal levels. Cells may be loaded with the AM ester forms of these calcium indicators by adding the dissolved indicator directly to dishes containing cultured cells. These indicators are useful for fluorescence and confocal microscopy, flow cytometry, and microplate screening applications.

Calcium Indicator (AM Ester) Specifications:
• Label (Ex/Em of Ca2+–bound form): Fluo-3 (506/526 nm)
• Fluorescence intensity increase upon binding Ca2+: >100 fold
• Kd for Ca2+ in buffer: ∼335 nM
• Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelength


Using TPEN to Control Heavy Metal Cations
In addition, BAPTA-based indicators such as these bind various heavy metal cations (e.g., Mn2+, Zn2+, Pb2+) with substantially higher affinity than Ca2+. Perturbations to calcium measurements caused by presence of these ions can be controlled using the heavy metal-selective chelator TPEN.

More Choices for Fluorescent Calcium Indicators
We offer a large selection of Molecular Probes™ calcium indicators for use in various experimental scenarios. For more information, review Fluorescent Ca2+ Indicators Excited with Visible Light—Section 19.3 in the Molecular Probes™ Handbook.

For UV-excitable Ca2+ indicators, protein-based Ca2+ indicators, conjugates of Ca2+ indicators, and for fluorescence-based indicators of other metal ions (i.e., Mg2+, Zn2+) review Indicators for Ca2+, Mg2+, Zn2+ and Other Metal Ions—Chapter 19 in the Molecular Probes™ Handbook.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Permeability
Cell-Permeant
Detection Method
Fluorescent
For Use With (Application)
Calcium Assay
For Use With (Equipment)
Confocal Microscope, Fluorescence Microscope, High Content Analysis Instrument, HTS Reader, Microplate Reader, Fluorescent Imager
Product Type
Stain
Dye Type
Fluorescent Dye-Based
Indicator
Calcium Indicator
Indicator Properties
Fluorescent Dye-Based
Quantity
20 x 50μg
Shipping Condition
Room Temperature

Contents & Storage

Store in freezer -5°C to -30°C and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text