Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

Phire Hot Start II PCR Master Mix

Catalog number: F125L
Have Questions?
Thermo Scientific™

Phire Hot Start II PCR Master Mix

Catalog number: F125L
Have Questions?
Catalog Number
F125L
Unit Size
1,000 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
200 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
F125L1,000 reactions
-
F125S200 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Thermo Scientific Phire Hot Start II PCR Master Mix is convenient 2X mix designed to minimize the number of pipetting steps. The master mix contains Phire Hot Start II DNA Polymerase, nucleotides and optimized reaction buffer including MgCl2. Only template and primers need to be added to PCR reaction.

Phire Hot Start II DNA Polymerase, inluded in the master mix, is an enhanced PCR enzyme for routine and high throughput PCR applications. It outperforms every Taq-based hot start polymerase on the market. Phire Hot Start II DNA Polymerase is significantly faster, extremely robust, and also capable of amplifying long DNA fragments with high yields. These features are achieved through advanced protein engineering of the polymerase. It incorporates a unique double-stranded DNA binding domain which allows short extension times (10 to 15 s/kb), improves yields, and increases fidelity 2-fold compared to Taq DNA polymerase (Cat. No. EP0072).

Highlights

  • Quick hot start—No reactivation step
  • Fast enzyme—Amplify four times faster than with hot start Taq
  • Robust—Minimal reaction optimization due to high inhibitor tolerance
  • High yields—Abundant products due to high efficiency
  • Longer PCR products—Amplify significantly longer DNA fragments than with any hot start Taq

Applications

  • Hot Start PCR
  • Routine PCR
  • Non-high fidelity PCR
  • Fast PCR
  • High throughput PCR
  • Genotyping

Note:The optimal annealing temperature for Phire DNA Polymerases may differ significantly from that of Taq-based polymerases. For optimal results start by accurately calculating your Tm with our Tm calculator.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

GC-Rich PCR Performance
Medium
Polymerase
Phire Hot Start II DNA Polymerase
Reaction Speed
Fast
Product Type
Hot Start PCR Master Mix
Shipping Condition
Wet or Dry Ice
For Use With (Application)
Cloning, Hot-start PCR
Concentration
2X
Fidelity (vs. Taq)
2X
Hot Start
Built-In Hot Start
No. of Reactions
1000 Reactions
Overhang
Blunt
Reaction Format
SuperMix or Master Mix
Starting Material
DNA

Contents & Storage

• 2X Phire Hot Start II PCR Master Mix, 20 x 1.25 mL
• DMSO, 4 x 0.5 mL
• Nuclease-free water, 20 x 1.25 mL

Store at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text