Invitrogen™

Fluorescein Diacetate (FDA)

Catalog number: F1303
Have Questions?
Invitrogen™

Fluorescein Diacetate (FDA)

Catalog number: F1303
Have Questions?
Catalog Number
F1303
Unit Size
1 g
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
F13031 g
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Fluorescein diacetate (FDA) is a cell-permeant esterase substrate that can serve as a viability probe that measures both enzymatic activity, which is require to activate its fluorescence, and cell-membrane integrity, which is required for intracellular retention of their fluorescent product. Upon hydrolysis by intracellular esterases, this AM ester yields fluorescein.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Fluorescein Diacetate
Dye Type
Classic Dyes
Reagent Type
Cell Tracker Compounds, Cell Labeling Reagents
Shipping Condition
Room Temperature

Contents & Storage

Store in freezer -5°C to -30°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text