Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

Phire Plant Direct PCR Kit (without sampling tools)

Catalog number: F130WH
Have Questions?
Thermo Scientific™

Phire Plant Direct PCR Kit (without sampling tools)

Catalog number: F130WH
Have Questions?
Catalog Number
F130WH
Unit Size
500 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
F130WH500 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Phire Plant Direct PCR Kit is designed to amplify DNA directly from plant samples. No DNA purification is required prior to PCR. The kit is based on Phire Hot Start II DNA Polymerase, a specially engineered enzyme with a DNA binding domain. The unique features of this DNA polymerase make it extremely robust and tolerant of many PCR inhibitors present in plant, fungi and bacteria.

The kit includes a complete set of optimized PCR reagents as well as detailed protocols for different sample types, making it an ideal choice for amplification of plant DNA. In addition, the kit includes control primers for positive control reactions.The Control Primers amplify a highly conserved region of chloroplast DNA and are universal for many plant species.

Highlights

• No need for time-consuming and expensive DNA purification steps
• Very little sample material required
• Two simple protocols for various applications
Phire Hot Start II DNA Polymerase provides high yields of specific product with short extension time (20 s/kb).

Applications

• Genotyping
• Transgene detection

The optimal annealing temperature for Phire DNA Polymerase may differ significantly from that of Taq -based polymerases. For optimal results start by accurately calculating your Tm with our Tm calculator.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

GC-Rich PCR Performance
Medium
PCR Method
Direct PCR
Polymerase
Phire Hot Start II DNA polymerase
Reaction Speed
Fast
Product Line
Phire
Product Type
Direct PCR Kit
Hot Start
Built-In Hot Start
No. of Reactions
500 x 20 μL Reactions
Overhang
Blunt
Reaction Format
Separate Components
Sample Type
Plant
Starting Material
DNA

Contents & Storage

Contains:
• 200 μL Phire Hot Start II DNA Polymerase
• 5 x 1.25 mL 2X Phire Plant Tissue PCR Buffer (includes dNTPs and MgCl2)
• 40 μL Universal Control Primer mix (25 μM each)
• 2 x 5 mL Dilution Buffer

Store at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text